Learn More
3-(N-Maleimidylpropionyl)biocytin, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(N-Maleimidylpropionyl)biocytin |
| CAS | 102849-12-7 |
| Molecular Formula | C23 H33 N5 O7 S |
| Quantity | 100 mg |
| Synonym | N2-[3-(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)-1-oxopropyl]-N6-[5-[(3aS,4S,6aR)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]-L-lysine |
| InChI Formula | InChI=1S/C23H33N5O7S/c29-17(7-2-1-6-16-21-15(13-36-16)26-23(35)27-21)24-11-4-3-5-14(22(33)34)25-18(30)10-12-28-19(31)8-9-20(28)32/h8-9,14-16,21H,1-7,10-13H2,(H,24,29)(H,25,30)(H,33,34)(H2,26,27,35)/t14-,15-,16-,21-/m0/s1 |
| SMILES | OC(=O)[C@H](CCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)NC(=O)CCN3C(=O)C=CC3=O |
| IUPAC Name | (2S)-6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-2-[3-(2,5-dioxopyrrol-1-yl)propanoylamino]hexanoic acid |
| Molecular Weight (g/mol) | 523.6 |
| Formula Weight | 523.2101 |