Learn More
3-n-Hexylthiophene, 98%
Product Code. 15492278
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492278 1 g 1g
15402288 5 g 5g
15412288 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15492278 Supplier Thermo Scientific Alfa Aesar Supplier No. H55891.03
Request Bulk or Custom Format
CAS: 1693-86-3 | C10H16S | 168.298 g/mol
3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.
Solubility
Partly soluble in water.
Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1693-86-3
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.298
MDL Number MFCD00143181
InChI Key JEDHEMYZURJGRQ-UHFFFAOYSA-N
Synonym 3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
PubChem CID 566849
IUPAC Name 3-hexylthiophene
SMILES CCCCCCC1=CSC=C1
Specifications
Density 0.936
Boiling Point 65°C (0.45 mmHg)
Flash Point 37°C (99°F)
Odor Stench
Refractive Index 1.496
Quantity 1 g
UN Number UN1993
Beilstein 1617129
Solubility Information Partly soluble in water.
Formula Weight 168.3
Percent Purity ≥99%
Chemical Name or Material 3-n-Hexylthiophene