missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene-2-carboxaldehyde, tech. 85%

Product Code. 11471708
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471708 25 g 25g
11481708 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471708 Supplier Thermo Scientific Alfa Aesar Supplier No. A13961.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5834-16-2 | C6H6OS | 126.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5834-16-2
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00005430
InChI Key BSQKBHXYEKVKMN-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarboxaldehyde, 3-methylthiophene-2-carboxaldehyde, 2-formyl-3-methylthiophene, 3-methylthiophene-2-aldehyde, 3-methyl-2-thiophenecarbaldehyde, 3-methyl-2-formylthiophene, 3-methyl-2-thiophene carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methyl, thiophenecarboxaldehyde, methyl, 3-methyl-thiophene-2-carbaldehyde
PubChem CID 79911
IUPAC Name 3-methylthiophene-2-carbaldehyde
SMILES CC1=C(SC=C1)C=O

Specifications

Density 1.17
Boiling Point 215°C
Flash Point 82°C (179°F)
Refractive Index 1.586
Quantity 25 g
Beilstein 107874
Sensitivity Air Sensitive
Formula Weight 126.18
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Methylthiophene-2-carboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.