Learn More
3-methylpentanal, TRC
Specifications
Specifications
| Chemical Name or Material | 3-methylpentanal |
| CAS | 15877-57-3 |
| Molecular Formula | C6H12O |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C6H12O/c1-3-6(2)4-5-7/h5-6H,3-4H2,1-2H3 |
| SMILES | CCC(C)CC=O |
| IUPAC Name | 3-methylpentanal |
| Molecular Weight (g/mol) | 100.15 |
| Formula Weight | 100.0888 |