missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyladamantan-1-amine Hydrochloride (Desmethylmemantine Hydrochloride), Mikromol™
Description
3-Methyladamantan-1-amine Hydrochloride (Desmethylmemantine Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Demethyl Memantine Hydrochloride |
| CAS | 33103-93-4 |
| Analyte or Component Names | Demethyl Memantine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H19 N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3-Methyladamantan-1-amine Hydrochloride, Tricyclo[3.3.1.13,7]decan-1-amine, 3-methyl-, hydrochloride (1:1), 1-Adamantanamine, 3-methyl-, hydrochloride (7CI,8CI), Tricyclo[3.3.1.13,7]decan-1-amine, 3-methyl-, hydrochloride, Demethyl Memantine Hydrochloride, Desmethylmemantine Hydrochloride, Memantine Related Compound G as Hydrochloride |
| InChI Formula | InChI=1S/C11H19N.ClH/c1-10-3-8-2-9(4-10)6-11(12,5-8)7-10;/h8-9H,2-7,12H2,1H3;1H |
| SMILES | Cl.CC12CC3CC(C1)CC(N)(C3)C2 |
| IUPAC Name | 3-methyladamantan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?