missing translation for 'onlineSavingsMsg'
Learn More

3-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10346804
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10346804

Brand: Thermo Scientific Maybridge CC76703DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 62522-63-8
Molecular Formula C8H6ClNO4S
Molecular Weight (g/mol) 247.65
MDL Number MFCD03033702
InChI Key MRSCMVKREGBNOW-UHFFFAOYSA-N
Synonym 3-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 3-methyl-2-oxo-2,3-dihydrobenzo d oxazole-6-sulfonyl chloride, 3-methyl-2-oxo-2,3-dihydro-benzooxazole-6-sulfonyl chloride, 6-benzoxazolesulfonyl chloride, 2,3-dihydro-3-methyl-2-oxo, 2,3-dihydro-3-methyl-2-oxo-6-benzoxazolesulfonyl chloride, 6-chlorosulfonyl-3-methylbenzoxazolin-2-one, 6-chlorosulfonyl-3-methyl-3-hydrobenzoxazol-2-one, pubchem5602, acmc-209n4g, 3-methyl-6-chlorosulfonylbenzoxazolone
PubChem CID 2772060
SMILES CN1C(=O)OC2=CC(=CC=C12)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 3-methyl-2-oxo-1,3-benzoxazole-6-sulfonyl chloride
Formula Weight 247.66
Percent Purity 97%
Chemical Name or Material 3-Methyl-2-oxo-2, 3-dihydro-1, 3-benzoxazole-6-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.