missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzyl alcohol, 99%

Product Code. 10307320 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10307320

Brand: Thermo Scientific Acros 182210050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 57455-06-8 | C7H7IO | 234.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 57455-06-8
CAS Min % 98.5
CAS Max % 100.0
Color Orange-Red to Yellow
Boiling Point 252°C (711.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7IO
Linear Formula IC6H4CH2OH
Refractive Index 1.6340 to 1.636
MDL Number MFCD00004635
Quantity 5 g
Synonym 3-iodophenyl methanol, 3-iodobenzyl alcohol, m-iodobenzyl alcohol, benzenemethanol, 3-iodo, benzyl alcohol, m-iodo, 3-iodo-benzyl alcohol, 3-iodobenzylalcohol, 3-iodo-phenyl-methanol, m-iodo benzylalcohol, 3-iodophenyl methan-1-ol
InChI Key QGCCNWSXJHGUNL-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)I)CO
IUPAC Name (3-iodophenyl)methanol
Molecular Weight (g/mol) 234.04
PubChem CID 42300
Formula Weight 234.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Iodobenzyl alcohol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Iodobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.