Learn More
3-Hydroxypropionic Acid Sodium Salt, TRC
Product Code. 30005367
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005367 10 g 10g
30003357 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30005367 Supplier Toronto Research Chemicals Supplier No. TRCH95248010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6487-38-3
Molecular Formula C3H5NaO3
Molecular Weight (g/mol) 112.06
Synonym Propanoic acid, 3-hydroxy-, sodium salt (1:1), Hydracrylic acid, sodium salt (6CI,8CI), Propanoic acid, 3-hydroxy-, monosodium salt (9CI), Sodium β-hydroxypropionate, Sodium 3-hydroxypropanoate, 3-Hydroxypropanoic acid sodium salt, 2-Deoxyglyceric acid sodium salt, 3-Hydroxypropionic acid sodium salt, Ethylenelactic acid sodium salt, β-Hydroxypropionic acid sodium salt, β-Lactic acid sodium salt
IUPAC Name sodium;3-hydroxypropanoate
SMILES [Na+].OCCC(=O)[O-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C3H6O3.Na/c4-2-1-3(5)6;/h4H,1-2H2,(H,5,6);/q;+1/p-1
Formula Weight 112.0136
Percent Purity >95%
Chemical Name or Material 3-Hydroxypropionic Acid Sodium
Grade HPLC