missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyhexadecanoylcarnitine Inner Salt (Mixture of Diastereomers), TRC

Product Code. 30012755
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012755 1 mg 1mg
30010466 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012755 Supplier Toronto Research Chemicals Supplier No. TRCH9430051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195207-76-2
Molecular Formula C23 H45 N O5
Molecular Weight (g/mol) 415.61
Synonym 1-Propanaminium, 3-carboxy-2-[(3-hydroxy-1-oxohexadecyl)oxy]-N,N,N-trimethyl-, inner salt, (2R)-, 1-Propanaminium, 3-carboxy-2-[(3-hydroxy-1-oxohexadecyl)oxy]-N,N,N-trimethyl-, inner salt, [1(R)]-, 3-Hydroxyhexadecanoylcarnitine, 3-Hydroxypalmitoylcarnitine, 3-Hydroxyhexadecanoyl L-carnitine, 3-Hydroxypalmitoyl L-carnitine
IUPAC Name (3R)-3-(3-hydroxyhexadecanoyloxy)-4-(trimethylazaniumyl)butanoate
SMILES CCCCCCCCCCCCCC(O)CC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C23H45NO5/c1-5-6-7-8-9-10-11-12-13-14-15-16-20(25)17-23(28)29-21(18-22(26)27)19-24(2,3)4/h20-21,25H,5-19H2,1-4H3/t20?,21-/m1/s1
Formula Weight 415.3298
Percent Purity >95%
Chemical Name or Material 3-Hydroxypalmitoyl L-Carnitine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.