missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy Carbamazepine tert-Butyldimethylsilyl Ether, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy Carbamazepine tert-Butyldimethylsilyl Ether |
| Analyte or Component Names | 3-Hydroxy Carbamazepine tert-Butyldimethylsilyl Ether |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 N2 O2 Si |
| InChI Formula | InChI=1S/C21H26N2O2Si/c1-21(2,3)26(4,5)25-17-13-12-16-11-10-15-8-6-7-9-18(15)23(20(22)24)19(16)14-17/h6-14H,1-5H3,(H2,22,24) |
| Solution Type | Neat |
| SMILES | CC(C)(C)[Si](C)(C)Oc1ccc2C=Cc3ccccc3N(C(=O)N)c2c1 |
| IUPAC Name | 2-[tert-butyl(dimethyl)silyl]oxybenzo[b][1]benzazepine-11-carboxamide |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 366.529 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?