missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy Carbamazepine tert-Butyldimethylsilyl Ether, TRC

Product Code. 30394225
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394225 1 mg 1mg
30404726 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394225 Supplier Toronto Research Chemicals Supplier No. TRCH8841151MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxy Carbamazepine tert-Butyldimethylsilyl Ether
Analyte or Component Names 3-Hydroxy Carbamazepine tert-Butyldimethylsilyl Ether
Shipping Condition Room Temperature
Molecular Formula C21 H26 N2 O2 Si
InChI Formula InChI=1S/C21H26N2O2Si/c1-21(2,3)26(4,5)25-17-13-12-16-11-10-15-8-6-7-9-18(15)23(20(22)24)19(16)14-17/h6-14H,1-5H3,(H2,22,24)
Solution Type Neat
SMILES CC(C)(C)[Si](C)(C)Oc1ccc2C=Cc3ccccc3N(C(=O)N)c2c1
IUPAC Name 2-[tert-butyl(dimethyl)silyl]oxybenzo[b][1]benzazepine-11-carboxamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 366.529
Quantity 1 mg
Formula Weight 366.176
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.