Learn More
3-Hydroxy-3-methyl-2-butanone oxime, 98%, Thermo Scientific™
Product Code. 11362906
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11362906 5 g 5g
11372906 25 g 25g
2 options
Product Code. 11362906
Brand: Thermo Scientific Alfa Aesar L10624.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7431-25-6
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00059656
InChI Key QKLLBCGVADVPKD-XQRVVYSFSA-N
Synonym 3-hydroxy-3-methyl-2-butanone oxime, 3e-3-hydroxyimino-2-methylbutan-2-ol, qkllbcgvadvpkd-xqrvvysfsa, 3-hydroxy-3-methyl-butan-2-one oxime, 3z-3-hydroxyimino-2-methylbutan-2-ol, 3e-3-hydroxyimino-2-methyl-butan-2-ol, 2z-3-hydroxy-3-methyl-2-butanone oxime #
PubChem CID 5385758
IUPAC Name (3Z)-3-hydroxyimino-2-methylbutan-2-ol
SMILES CC(=NO)C(C)(C)O
Specifications
Melting Point 84°C to 86°C
Quantity 5 g
Beilstein 1747660
Formula Weight 117.15
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone oxime