Learn More
3-Heptyne, 97%
Product Code. p-7035777
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440398 5 g 5g
15450398 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15440398 Supplier Thermo Scientific Alfa Aesar Supplier No. H53504.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 2586-89-2 | C7H12 | 96.173 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2586-89-2
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00039960
InChI Key KLYHSJRCIZOUHE-UHFFFAOYSA-N
Synonym 3-heptyne, 1-ethyl-2-propylacetylene, acmc-20an2z
PubChem CID 75755
IUPAC Name hept-3-yne
SMILES CCCC#CCC
Specifications
Density 0.741
Boiling Point 105°C to 106°C
Flash Point −16°C (−3°F)
Linear Formula CH3(CH2)2C≡CCH2CH3
Refractive Index 1.4225
Quantity 5 g
UN Number UN3295
Beilstein 1731759
Formula Weight 96.17
Percent Purity 97%
Chemical Name or Material 3-Heptyne