missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorothiophenol, 98%

Product Code. p-7026850
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11476326

Brand: Thermo Scientific Alfa Aesar B23114.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2557-77-9 | C6H5FS | 128.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2557-77-9
Molecular Formula C6H5FS
Molecular Weight (g/mol) 128.16
MDL Number MFCD00040227
InChI Key ZDEUGINAVLMAET-UHFFFAOYSA-N
Synonym 3-fluorothiophenol, m-fluorothiophenol, m-fluorobenzenethiol, 3-fluoro thiophenol, benzenethiol, 3-fluoro, 3-fluorobenzene-1-thiol, 1-fluoro-3-mercaptobenzene, 3-fluoro-thiophenol, 3-fluoro benzenethiol, 3-fluoro-benzenethiol
PubChem CID 75710
IUPAC Name 3-fluorobenzenethiol
SMILES FC1=CC=CC(S)=C1

Specifications

Density 1.203
Boiling Point 156°C
Flash Point 58°C (136°F)
Odor Stench
Refractive Index 1.554
Quantity 1 g
UN Number UN3071
Beilstein 2039302
Sensitivity Air Sensitive
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material 3-Fluorothiophenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.