missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxypropylamine, 99%

Artikelnummer. p-7027155
Change view
Click to view available options
Quantity:
100 g
500 g
Packungsgröße:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11430108 100 g 100g
11440108 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11430108 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. B23694.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 6291-85-6 | C5H13NO | 103.165 g/mol

It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.

Solubility
Miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 6291-85-6
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00008221
InChI Key SOYBEXQHNURCGE-UHFFFAOYSA-N
Synonym 3-ethoxypropylamine, 1-propanamine, 3-ethoxy, 3-ethoxy-1-propanamine, 3-ethoxy-1-propylamine, propylamine, 3-ethoxy, 3-ethoxy-propylamine, 3-aminopropyl ethyl ether, 3-ethyoxypropylamine, 3-ethoxypropyl amine, 3-ethyloxy-propylamine
PubChem CID 22720
IUPAC Name 3-ethoxypropan-1-amine
SMILES CCOCCCN

Spezifikation

Boiling Point 136°C to 138°C
Flash Point 32°C (91°F)
Linear Formula CH3CH2O(CH2)3NH2
Quantity 100 g
UN Number UN2733
Beilstein 1732287
Formula Weight 103.16
Percent Purity 99%
Chemical Name or Material 3-Ethoxypropylamine
compliance-icons compliance-icons
Produktidentifikator
  • 3-Ethoxypropylamine
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Ergänzende Informationen
  • MIXTURE LIST-Contains : 3-ethoxy-1-Propanamine

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.