missing translation for 'onlineSavingsMsg'
Learn More

3-Deazaadenosine hydrochloride, MedChemExpress

Product Code. 16442688
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442688 5 mg 5mg
16473439 1 mg 1mg
16430479 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16442688 Supplier Medchem Express Supplier No. HYW013332A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Deazaadenosine (hydrochloride) is an inhibitor of S-adenosylhomocysteine hydrolase, with a Ki of 3.9 μM; 3-Deazaadenosine has anti-inflammatory, anti-proliferative and anti-HIV activity.

3-Deazaadenosine (hydrochloride) is an inhibitor of S-adenosylhomocysteine hydrolase, with a Ki of 3.9 μM

Specifications

Chemical Name or Material 3-Deazaadenosine hydrochloride
CAS 86583-19-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H15ClN4O4
Quantity 5 mg
Solubility Information DMSO : 41.67 mg/mL (137.66 mM; Need ultrasonic)
SMILES O[C@H]1[C@@H](O[C@H](CO)[C@H]1O)N2C3=C(C(N)=NC=C3)N=C2.Cl
Molecular Weight (g/mol) 302.71
Formula Weight 302.71
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.