missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15417237
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15417237

Brand: Thermo Scientific Alfa Aesar H34296.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1240257-14-0
Molecular Formula C7H5Br2F5S
Molecular Weight (g/mol) 375.977
MDL Number MFCD16652484
InChI Key PSRNYAZILJJJGP-UHFFFAOYSA-N
Synonym 3-bromo-5-pentafluorosulfur benzyl bromide, 3-bromo-5-pentafluorothio benzyl bromide, 1-bromo-3-bromomethyl-5-pentafluoro-??-sulfanyl benzene
PubChem CID 66523602
IUPAC Name [3-bromo-5-(bromomethyl)phenyl]-pentafluoro-$l^{6}-sulfane
SMILES C1=C(C=C(C=C1S(F)(F)(F)(F)F)Br)CBr

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 375.98
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(pentafluorothio)benzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.