missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chloropyridine, 97%

Product Code. 11906381 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11906381

Brand: Thermo Scientific Acros 437400050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73583-39-8 | C5H3BrClN | 192.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73583-39-8
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
InChI Key BELDOPUBSLPBCQ-UHFFFAOYSA-N
Synonym 3-chloro-5-bromopyridine, 3-bromo-5-chloro-pyridine, pyridine, 3-bromo-5-chloro, 5-bromo-3-chloropyridine, pubchem7700, 3-brom-5-chlorpyridin, acmc-209orf, #, ksc377a4j, 3-bromo-5-chloropyridine
PubChem CID 606256
IUPAC Name 3-bromo-5-chloropyridine
SMILES C1=C(C=NC=C1Br)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 78.7°C to 80.1°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 192.44
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-chloropyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromo-5-chloropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.