Learn More
3-(Benzyloxy)-2,2-dimethyl-1-propanol, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Benzyloxy)-2,2-dimethyl-1-propanol |
| CAS | 66582-32-9 |
| Molecular Formula | C12H18O2 |
| Quantity | 250 mg |
| Synonym | 2,2-Dimethyl-3-(phenylmethoxy)-1-propanol |
| InChI Formula | InChI=1S/C12H18O2/c1-12(2,9-13)10-14-8-11-6-4-3-5-7-11/h3-7,13H,8-10H2,1-2H3 |
| SMILES | CC(C)(CO)COCc1ccccc1 |
| IUPAC Name | 2,2-dimethyl-3-phenylmethoxypropan-1-ol |
| Molecular Weight (g/mol) | 194.27 |
| Formula Weight | 194.1307 |