missing translation for 'onlineSavingsMsg'
Learn More

3'-Azido-3'-deoxythymidine Beta-D-glucuronide, Sodium Salt, TRC

Product Code. 30133494
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133494 25 mg 25mg
30009675 10 mg 10mg
30006486 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133494 Supplier Toronto Research Chemicals Supplier No. TRCA82505025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133525-01-6
Molecular Formula C16 H20 N5 O10 . Na
Molecular Weight (g/mol) 465.35
Synonym Thymidine, 3'-azido-3'-deoxy-5'-O-β-D-glucopyranuronosyl-, monosodium salt (9CI), 3'-Azido-3'-deoxythymidine-5'-O-β-D-glucuronide sodium salt, 3'-Azido-3'-deoxythymidine-β-D-glucuronide sodium salt, 3'-Azido-3'-deoxy-5'-O-β-D-glucopyranuronosylthymidine sodium salt, 3'-Deoxy-3'-azidothymidine-5'-β-glucuronide sodium salt, AZT 5'-glucuronide sodium salt, GAZT sodium, Glo-azT Na, Retrovir glucuronide sodium, Zidovudine 5'-glucuronide sodium salt, Zidovudine glucuronide sodium salt
IUPAC Name sodium;(2S,3S,4S,5R,6R)-6-[[(2S,3S,5R)-3-azido-5-(5-methyl-2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methoxy]-3,4,5-trihydroxyoxane-2-carboxylate
SMILES [Na+].CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)[O-])O2)C(=O)NC1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H21N5O10.Na/c1-5-3-21(16(28)18-13(5)25)8-2-6(19-20-17)7(30-8)4-29-15-11(24)9(22)10(23)12(31-15)14(26)27;/h3,6-12,15,22-24H,2,4H2,1H3,(H,26,27)(H,18,25,28);/q;+1/p-1/t6-,7+,8+,9-,10-,11+,12-,15+;/m0./s1
Formula Weight 465.1108
Percent Purity >95%
Chemical Name or Material 3'-Azido-3'-deoxythymidine beta-D-glucuronide, Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.