Learn More
3-(Aminooxy)-1-propanethiol Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Aminooxy)-1-propanethiol Hydrochloride |
| CAS | 1071-99-4 |
| Molecular Formula | C3 H9 N O S . Cl H |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C3H9NOS.ClH/c4-5-2-1-3-6;/h6H,1-4H2;1H |
| SMILES | Cl.NOCCCS |
| IUPAC Name | O-(3-sulfanylpropyl)hydroxylamine;hydrochloride |
| Molecular Weight (g/mol) | 143.64 |
| Formula Weight | 143.0172 |