Learn More
3-Amino-2-methylpropanoic acid, MedChemExpress
Product Code. 16459366
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16459366
Brand: Medchem Express HYW012974
MedChemExpress 3-Amino-2-methylpropanoic acid could induce browning of white fat and hepatic β-oxidation and is inversely correlated with cardiometabolic risk factors.
3-Amino-2-methylpropanoic acid could induce browning of white fat and hepatic β-oxidation and is inversely correlated with cardiometabolic risk factors.
Specifications
Chemical Name or Material 3-Amino-2-methylpropanoic acid
CAS 144-90-1
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C4H9NO2
Quantity 100 mg
Solubility Information H2O : 100 mg/mL (969.74 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (9.70 mM; Need ultrasonic)
SMILES O=C(O)C(C)CN
Molecular Weight (g/mol) 103.12
Formula Weight 103.12
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation