Learn More
3,7-Dimethyl-2-octen-1-ol (cis trans mixture), TRC
Specifications
Specifications
| Chemical Name or Material | 3,7-Dimethyl-2-octen-1-ol (cis trans mixture) |
| CAS | 40607-48-5 |
| Molecular Formula | C10H20O |
| Quantity | 250 mg |
| SMILES | OC/C=C(C)/CCCC(C)C |
| Molecular Weight (g/mol) | 156.27 |