missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichlorophthalic Anhydride, TRC

Product Code. 30388677
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388677 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388677 Supplier Toronto Research Chemicals Supplier No. TRCD4344902.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,6-Dichlorophthalic Anhydride
CAS 4466-59-5
Molecular Formula C8 H2 Cl2 O3
Quantity 2.5 g
Synonym 4,7-Dichloroisobenzofuran-1,3-dione, 3,6-Dichlorophthalic Acid Anhydride, 3,6-Dichlorophthalic Anhydride, 4,7-Dichloro-1,3-isobenzofurandione
InChI Formula InChI=1S/C8H2Cl2O3/c9-3-1-2-4(10)6-5(3)7(11)13-8(6)12/h1-2H
SMILES Clc1ccc(Cl)c2C(=O)OC(=O)c12
IUPAC Name 4,7-dichloroisobenzofuran-1,3-dione
Molecular Weight (g/mol) 217.006
Formula Weight 215.9381
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.