Learn More
3,6-Di-O-(Alpha-D-mannopyranosyl)-D-mannopyrannose, TRC
Product Code. 30382996
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382996 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30382996 Supplier Toronto Research Chemicals Supplier No. TRCD46065010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,6-Di-O-(a-D-mannopyranosyl)-D-mannopyrannose
CAS 121123-33-9
Molecular Formula C18 H32 O16
Quantity 10 mg
InChI Formula InChI=1S/C18H32O16/c19-1-4-7(21)10(24)12(26)17(32-4)30-3-6-9(23)15(14(28)16(29)31-6)34-18-13(27)11(25)8(22)5(2-20)33-18/h4-29H,1-3H2/t4-,5-,6-,7-,8-,9-,10+,11+,12+,13+,14+,15+,16?,17+,18-/m1/s1
SMILES OC[C@H]1O[C@H](OC[C@H]2OC(O)[C@@H](O)[C@@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]3O)[C@@H]2O)[C@@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-[[(2R,3R,4S,5S)-3,5,6-trihydroxy-4-[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]methoxy]oxane-3,4,5-triol
Molecular Weight (g/mol) 504.44
Formula Weight 504.169