missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzaldehyde, 98%

Product Code. 11342247
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342247 1 g 1g
11352247 5 g 5g
11362247 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342247 Supplier Thermo Scientific Alfa Aesar Supplier No. L09748.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5779-95-3 | C9H10O | 134.178 g/mol

3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.

Solubility
Not miscible with water.

Notes
Handle and store under inert gas. Air Sensitive. Store away from air and oxidizing agents. Incompatible with oxidizing agents and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5779-95-3
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00082777
InChI Key NBEFMISJJNGCIZ-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dimethyl, 3,5-dimethyl benzaldehyde, m-xylene-5-carboxaldehyde, 3,5-dimethyl-benzaldehyde, pubchem15515, acmc-1arkp, 3,5-dimehtylbenzaldehyde, #, ksc494c0b, 3,5-dimethylbenzaldehyde
PubChem CID 34225
IUPAC Name 3,5-dimethylbenzaldehyde
SMILES CC1=CC(=CC(=C1)C=O)C

Specifications

Melting Point 8°C to 10°C
Boiling Point 220°C to 222°C
Refractive Index 1.5385
Quantity 1 g
Beilstein 2038840
Sensitivity Air Sensitive
Solubility Information Not miscible with water.
Formula Weight 134.18
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethylbenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.