missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Diaminobenzotrifluoride, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3,5-Diaminobenzotrifluoride |
| CAS | 368-53-6 |
| Molecular Formula | C7H7N2F3 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C7H7F3N2/c8-7(9,10)4-1-5(11)3-6(12)2-4/h1-3H,11-12H2 |
| SMILES | Nc1cc(N)cc(c1)C(F)(F)F |
| IUPAC Name | 5-(trifluoromethyl)benzene-1,3-diamine |
| Molecular Weight (g/mol) | 176.13 |
| Formula Weight | 176.0561 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction