missing translation for 'onlineSavingsMsg'
Learn More

3’, 5’-Di-O-acetyl-5-bromo-2’-deoxyuridine, TRC

Product Code. 30351813
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351813 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351813 Supplier Toronto Research Chemicals Supplier No. TRCD3094002G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3', 5'-Di-O-acetyl-5-bromo-2'-deoxyuridine
CAS 6161-23-5
Molecular Formula C13 H15 Br N2 O7
Quantity 2 g
InChI Formula InChI=1S/C13H15BrN2O7/c1-6(17)21-5-10-9(22-7(2)18)3-11(23-10)16-4-8(14)12(19)15-13(16)20/h4,9-11H,3,5H2,1-2H3,(H,15,19,20)/t9-,10+,11+/m0/s1
SMILES CC(=O)OC[C@H]1O[C@H](C[C@@H]1OC(=O)C)N2C=C(Br)C(=O)NC2=O
IUPAC Name [(2R,3S,5R)-3-acetyloxy-5-(5-bromo-2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol) 391.17
Formula Weight 390.0063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.