Learn More
3,4-Lutidine, 98+%
Product Code. 11352176
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352176 25 mL 25mL
11362176 100 mL 100mL
11372176 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11352176 Supplier Thermo Scientific Alfa Aesar Supplier No. L03489.AC
Request Bulk or Custom Format
CAS: 583-58-4 | C7H9N | 107.156 g/mol
3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.
Solubility
Miscible with water, ethanol and ether.
Notes
Hygroscopic. Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 583-58-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006403
InChI Key NURQLCJSMXZBPC-UHFFFAOYSA-N
PubChem CID 11417
IUPAC Name 3,4-dimethylpyridine
SMILES CC1=C(C=NC=C1)C
Specifications
Melting Point -11°C
Density 0.955
Boiling Point 163°C to 164°C
Flash Point 47°C (116°F)
Odor Unpleasant
Refractive Index 1.512
Quantity 25 mL
UN Number UN2929
Beilstein 106583
Synonym 3,4-lutidine, pyridine, 3,4-dimethyl, unii-fcu895kj7h, pyridine,3,4-dimethyl, fcu895kj7h, 3,4-lutidene, t 3,4-lutidine, 3,4-dimethyl-pyridine, 5-20-06-00054 beilstein handbook reference
Solubility Information Miscible with water,ethanol and ether.
Formula Weight 107.16
Percent Purity ≥98%
Chemical Name or Material 3,4-Lutidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4-Lutidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3,4-Lutidine