Learn More
3,4-Dichloro-6-aminotoluene, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-Dichloro-6-aminotoluene |
| CAS | 2387-08-8 |
| Molecular Formula | C7H7Cl2N |
| Quantity | 500 mg |
| Synonym | 4,5-Dichloro-2-aminotoluene, 4,5-Dichloro-2-methylaniline, 4,5-Dichloro-o-toluidine |
| InChI Formula | InChI=1S/C7H7Cl2N/c1-4-2-5(8)6(9)3-7(4)10/h2-3H,10H2,1H3 |
| SMILES | ClC1=C(Cl)C=C(C)C(N)=C1 |
| IUPAC Name | 4,5-dichloro-2-methylaniline |
| Molecular Weight (g/mol) | 176.04 |
| Formula Weight | 175 |