Learn More
MedChemExpress 3,3'-Dioctadecyloxacarbocyanine perchlorate
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | DMF : 10 mg/mL (11.34 mM; Need ultrasonic) ∣ DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Target | HBV |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 881.7 g/mol |
| CAS | 34215-57-1 |
| Synonym | DiO ∣ DiOC18(3) |