Learn More
(3,3-Dimethyloxiran-2-yl)methanol (>85%), TRC
Specifications
Specifications
| Chemical Name or Material | (3,3-Dimethyloxiran-2-yl)methanol |
| CAS | 18511-56-3 |
| Molecular Formula | C5H10O2 |
| Quantity | 25 mg |
| Synonym | (3,3-Dimethyloxiranyl) Methanol |
| InChI Formula | InChI=1S/C5H10O2/c1-5(2)4(3-6)7-5/h4,6H,3H2,1-2H3 |
| SMILES | CC1(C(O1)CO)C |
| IUPAC Name | (3,3-dimethyloxiran-2-yl)methanol |
| Molecular Weight (g/mol) | 102.13 |