Learn More
3,3-Dimethylacrylic acid, 98%
Product Code. 11379539
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11379539
Brand: Thermo Scientific Alfa Aesar A16865.14
Request Bulk or Custom Format
CAS: 541-47-9 | C5H8O2 | 100.117 g/mol
Chemical Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chemical Intermediates.
Solubility
Soluble in water, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 541-47-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00004366
InChI Key YYPNJNDODFVZLE-UHFFFAOYSA-N
Synonym 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid
PubChem CID 10931
ChEBI CHEBI:37127
IUPAC Name 3-methylbut-2-enoic acid
SMILES CC(=CC(=O)O)C
Specifications
Melting Point 66°C to 68°C
Density 0.969
Boiling Point 198°C to 200°C
Flash Point 93°C (199°F)
Quantity 25 g
UN Number UN3261
Beilstein 1720305
Solubility Information Soluble in water,and methanol.
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material 3,3-Dimethylacrylic acid