Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass Stopcocks
Click to view available options
Plug Bore:
2.5 mm
4 mm
Description:
Plug size: NS 12.5
Plug size: NS 14.5
Plug size: NS 18.8
Includes:
PTFE Plug, Screw Thread Retaining Device
Solid Glass Plug
Solid Glass Plug, Capillary Side Arms
Unit Size:
Each
Pack of 10
Specifications
Specifications
| Type | Stopcock |
| Plug Bore | 4 mm |
| Description | Plug size: NS 14.5 |
| Material | Borosilicate Glass |
| Includes | Solid Glass Plug |