Learn More
3-(2-Aminoethyl)pyridine, 98%
Product Code. 11302987
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302987 5 g 5g
11392977 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11302987 Supplier Thermo Scientific Alfa Aesar Supplier No. L16311.06
Request Bulk or Custom Format
CAS: 20173-24-4 | C7H11N2 | 123.18 g/mol
3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20173-24-4
Molecular Formula C7H11N2
Molecular Weight (g/mol) 123.18
MDL Number MFCD00191604
InChI Key NAHHNSMHYCLMON-UHFFFAOYSA-O
Synonym 3-2-aminoethyl pyridine, 2-pyridin-3-yl ethanamine, 3-aminoethylpyridine, 3-pyridineethanamine, 2-pyridin-3-yl ethan-1-amine, 2-3-pyridyl ethylamine, 3-pyridineethaneamine, 3-pyridine ethanamine, 2-pyridin-3-yl-ethylamine, 2-3-pyridinyl ethanamine
PubChem CID 854051
SMILES [NH3+]CCC1=CC=CN=C1
Specifications
Boiling Point 119°C to 120°C (19 mmHg)
Odor Amine-like
Refractive Index 1.5518
Quantity 5 g
UN Number UN2735
Beilstein 111288
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
IUPAC Name 2-pyridin-3-ylethanamine
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 3-(2-Aminoethyl)pyridine