missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Amino-2-oxoethyl)-5-methylhexanoic Acid, TRC

Product Code. 30323273
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323273 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323273 Supplier Toronto Research Chemicals Supplier No. TRCA61904550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2-Amino-2-oxoethyl)-5-methylhexanoic Acid
CAS 181289-15-6
Purity Grade Notes HPLC
Molecular Formula C9 H17 N O3
Quantity 50 g
Synonym 3-(Carbamoylmethyl)-5-methylhexanoic Acid
InChI Formula InChI=1S/C9H17NO3/c1-6(2)3-7(4-8(10)11)5-9(12)13/h6-7H,3-5H2,1-2H3,(H2,10,11)(H,12,13)
SMILES CC(C)CC(CC(=O)N)CC(=O)O
IUPAC Name 3-(2-amino-2-oxo-ethyl)-5-methyl-hexanoic acid
Molecular Weight (g/mol) 187.24
Formula Weight 187.1208
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.