missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Adamantyl)-4-methoxybenzeneboronic acid, 96%, Thermo Scientific™

Product Code. 15420386
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420386 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15420386 Supplier Thermo Scientific Alfa Aesar Supplier No. H53104.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Promotes the ortho hydroxalkylation of phenols by aldehyde. Used as reagent in the proection of thiols as cyclic boronates. This has been utilized to trap cis thiols in oder to prevent over oxidation during dihydroxylation reaction
TRUSTED_SUSTAINABILITY

Specifications

CAS 459423-32-6
Melting Point 239°C to 242°C
Molecular Formula C17H23BO3
MDL Number MFCD08056355
Quantity 250 mg
Synonym 3-1-adamantyl-4-methoxyphenylboronic acid, 3-adamantan-1-yl-4-methoxyphenyl boronic acid, 3-adamantan-1-yl-4-methoxyphenylboronic acid, boronic acid, 4-methoxy-3-tricyclo 3.3.1.13,7 dec-1-ylphenyl, 3-1-adamantyl-4-methoxyphenyl boronic acid, 3-1-adamantyl-4-methoxy-phenyl boronic acid, 3-adamantan-1-yl-4-methoxybenzeneboronic acid, 4-methoxy-3-3r,5s,7s-adamantan-1-yl phenylboronic acid, 4-methoxy-3-tricyclo 3.3.1.1∼3,7∼ decan-1-yl phenyl boronic acid
InChI Key FPZOBRFHRPQGAA-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)OC)C23CC4CC(C2)CC(C4)C3)(O)O
IUPAC Name [3-(1-adamantyl)-4-methoxyphenyl]boronic acid
Molecular Weight (g/mol) 286.178
PubChem CID 23148592
Formula Weight 286.18
Percent Purity 96%
Chemical Name or Material 3-(1-Adamantyl)-4-methoxybenzeneboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.