missing translation for 'onlineSavingsMsg'
Learn More

(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane, 97%, Thermo Scientific™

Product Code. p-4228633 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11975951

Brand: Thermo Scientific Acros 435171000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 695816-47-8
Molecular Formula C15H21O2P
Molecular Weight (g/mol) 264.3
InChI Key JAKXTLMNIHPLEU-RYUDHWBXSA-N
Synonym 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min
PubChem CID 16218307
IUPAC Name 2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane
SMILES CC1CCC(P1C2=CC=CC=C2C3OCCO3)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1030g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Specific Gravity 1.103
Formula Weight 264.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.