missing translation for 'onlineSavingsMsg'
Learn More

(2S,3R)-N-(tert-Butyloxycarbonyl)-O-(tert-butyl)dimethylsilyl-3,4-dehydro-3-methyl-pyroglutaminol, TRC

Product Code. 30347551
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347551 10 mg 10mg
30343902 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30347551 Supplier Toronto Research Chemicals Supplier No. TRCB80705010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,3R)-N-(tert-Butyloxycarbonyl)-O-(tert-butyl)dimethylsilyl-3,4-dehydro-3-methyl-pyroglutaminol
Analyte or Component Names (2S,3R)-N-(tert-Butyloxycarbonyl)-O-(tert-butyl)dimethylsilyl-3,4-dehydro-3-methyl-pyroglutaminol
Shipping Condition Room Temperature
Molecular Formula C17 H31 N O4 Si
InChI Formula InChI=1S/C17H31NO4Si/c1-12-10-14(19)18(15(20)22-16(2,3)4)13(12)11-21-23(8,9)17(5,6)7/h10,13H,11H2,1-9H3/t13-/m1/s1
Solution Type Neat
SMILES CC1=CC(=O)N([C@@H]1CO[Si](C)(C)C(C)(C)C)C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2S)-2-[[tert-butyl(dimethyl)silyl]oxymethyl]-3-methyl-5-oxo-2H-pyrrole-1-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 341.518
Quantity 10 mg
Formula Weight 341.202
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.