missing translation for 'onlineSavingsMsg'
Learn More

(2S,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Pilocarpic Acid Sodium Salt), Mikromol™

Product Code. 18040800
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18040800 25 mg 25mg
18090510 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18040800 Supplier Mikromol Supplier No. MM0338.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Pilocarpic Acid Sodium Salt)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Pilocarpic Acid Sodium Salt)
CAS 92598-79-3
Analyte or Component Names (2S,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Pilocarpic Acid Sodium Salt)
Shipping Condition Room Temperature
Molecular Formula C11 H17 N2 O3 . Na
Certifications/Compliance ISO 17025
Synonym Pilocarpine Hydrochloride Imp. B (EP), Pilocarpine Imp. B (EP), (2S,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt, Pilocarpine Hydrochloride Imp. B (EP) as Sodium Salt, Pilocarpic Acid Sodium Salt, 1H-Imidazole-5-butanoic acid, alpha-ethyl-beta-(hydroxymethyl)-1-methyl-, monosodium salt, [S-(R*,S*)]- (9CI), Sodium pilocarpate, Pilocarpine Hydrochloride Impurity B as Sodium Salt, Pilocarpine Nitrate Impurity B as Sodium Salt, Pilocarpine Impurity B as Sodium Salt, Pilocarpine Impurity B as Sodium Salt
InChI Formula InChI=1S/C11H18N2O3.Na/c1-3-10(11(15)16)8(6-14)4-9-5-12-7-13(9)2;/h5,7-8,10,14H,3-4,6H2,1-2H3,(H,15,16);/q;+1/p-1/t8-,10-;/m0./s1
SMILES [Na+].CC[C@@H]([C@H](CO)Cc1cncn1C)C(=O)[O-]
IUPAC Name sodium;(2S,3R)-2-ethyl-3-(hydroxymethyl)-4-(3-methylimidazol-4-yl)butanoate
Molecular Weight (g/mol) 248.25
Quantity 25 mg
Formula Weight 248.1137 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.