missing translation for 'onlineSavingsMsg'
Learn More

(2E)-Dehydrodonepezil (Donepezil Impurity), TRC

Product Code. 30134713
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134713 100 mg 100mg
30007006 50 mg 50mg
30006057 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134713 Supplier Toronto Research Chemicals Supplier No. TRCD229775100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145546-80-1
Molecular Formula C24H27NO3
Molecular Weight (g/mol) 377.48
Synonym 1H-Inden-1-one, 2,3-dihydro-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methylene]-, (2E)-, 1H-Inden-1-one, 2,3-dihydro-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methylene]-, (E)-, (E)-2-[(1-Benzylpiperidin-4-yl)methylene]-5,6-dimethoxyindan-1-one, (2E)-Dehydrodonepezil
IUPAC Name (2E)-2-[(1-benzylpiperidin-4-yl)methylidene]-5,6-dimethoxy-3H-inden-1-one
SMILES COc1cc2C\C(=C/C3CCN(Cc4ccccc4)CC3)\C(=O)c2cc1OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H27NO3/c1-27-22-14-19-13-20(24(26)21(19)15-23(22)28-2)12-17-8-10-25(11-9-17)16-18-6-4-3-5-7-18/h3-7,12,14-15,17H,8-11,13,16H2,1-2H3/b20-12+
Formula Weight 377.1991
Percent Purity >95%
Chemical Name or Material (E)-2-[(1-Benzylpiperidin-4-yl)methylene]-5,6-dimethoxyindan-1-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.