missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linezolid N-Oxide, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Linezolid N-Oxide
Specifications
Specifications
| Chemical Name or Material | Linezolid N-Oxide |
| CAS | 189038-36-6 |
| Analyte or Component Names | Linezolid N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H20 F N3 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-[[(5S)-3-[3-fluoro-4-(4-oxido-4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]-acetamide, Acetamide, N-[[3-[3-fluoro-4-(4-oxido-4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]-, (S)- (9CI), Linezolid N-Oxide |
| InChI Formula | InChI=1S/C16H20FN3O5/c1-11(21)18-9-13-10-19(16(22)25-13)12-2-3-15(14(17)8-12)20(23)4-6-24-7-5-20/h2-3,8,13H,4-7,9-10H2,1H3,(H,18,21)/t13-/m0/s1 |
| SMILES | CC(=O)NC[C@H]1CN(C(=O)O1)c2ccc(c(F)c2)[N+]3([O-])CCOCC3 |
| IUPAC Name | N-[[(5S)-3-[3-fluoro-4-(4-oxidomorpholin-4-ium-4-yl)phenyl]-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction