Learn More
Ethanol 96% +DEP 25L, Solveco
Description
Ethanol with an alcohol content of 96%, denatured using DEP (diethyl phthalate). The denaturing process is completely odorless, making this ethanol especially suitable for use in the formulation of perfumes, skin care products, and other cosmetic applications where fragrance neutrality is essential.
The ethanol is derived from natural, renewable agricultural sources and is free from added fragrances or impurities, ensuring compatibility with high-quality formulations.
A version with 99% alcohol content is also available for applications requiring lower water content or increased volatility.
Specifications
Specifications
| Chemical Name or Material | Ethyl Alcohol |
| CAS | 64-17-5 |
| Boiling Point | 78°C |
| Quantity | 25 L |
| Packaging | 15 L HDPE Container |
| Concentration | 96% |
| Molecular Weight (g/mol) | 46.07 g/mol |
| Percent Purity | 96% |
| Grade | Technical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.