Learn More
SI Analytics™ 250ML Electrolyte 3.5 MOL / L 5 mol / l, sterilized
Product Code. 11974739 Shop All SI Analytics Products
Click to view available options
Capacity (Metric):
0.8 kg
Unit Size:
250mL
Product Code. 11974739
Brand: SI Analytics™ 285138127
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Robust, reliable, ready-to-use 250mL electrolyte 3.5 MOL.
  • 250mL electrolyte 3.5 MOL / L 5 mol / l, sterilized
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 0.8 kg
Description 250ML Electrolyte 3.5 MOL
Type pH Electrode Accessories