missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxymetholone, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Oxymetholone
Specifications
Specifications
| Chemical Name or Material | Oxymetholone |
| CAS | 434-07-1 |
| Analyte or Component Names | Oxymetholone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H32 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 17-Beta-Hydroxy-2-hydroxymethylene-17-alpha-methyl-3-androstanone, 2-(Hydroxymethylene)-17-methyldihydrotestosterone, Adroyd, Anadrol, Anapolan 50, Anapolon, Anasteron, Anasteronal, Anasterone, Becorel, C.I. 406, HMD, NSC-26198, Nastenon, Oxymethenolone, Oxymetholone, Pardroyd, Plenastril, Protanabol, Roboral, Synasteron, Synasteron 50, Androstan-3-one, 17-hydroxy-2-(hydroxymethylene)-17-methyl-, (5alpha,17beta)-, 5alpha-Androstan-3-one, 17beta-hydroxy-2-(hydroxymethylene)-17-methyl- (6CI,8CI), 17alpha-Methyl-2-hydroxymethylene-17-hydroxy-5alpha-androstan-3-one, 17beta-Hydroxy-2-(hydroxymethylene)-17-methyl-5alpha-androstan-3-one, 17beta-Hydroxy-2-(hydroxymethylene)-17alpha-methyl-5alpha-androstan-3-one, 2-Hydroxymethylene-17alpha-methyl-17beta-hydroxy-3-androstanone, 2-Hydroxymethylene-17alpha-methylandrostan-17beta-ol-3-one, 2-Hydroxymethylene-17beta-hydroxy-17alpha-methyl-5alpha-androstan-3-one, Stanozolol Impurity B |
| InChI Formula | InChI=1S/C21H32O3/c1-19-11-13(12-22)18(23)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,22,24H,4-11H2,1-3H3/b13-12-/t14-,15+,16-,17-,19-,20-,21-/m0/s1 |
| SMILES | C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4CC(=O)\C(=C/O)\C[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (2Z,5S,8R,9S,10S,13S,14S,17S)-17-hydroxy-2-(hydroxymethylidene)-10,13,17-trimethyl-1,4,5,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction