missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylammonium Salicylate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Diethylammonium Salicylate
Specifications
Specifications
| Chemical Name or Material | Diethylammonium Salicylate |
| CAS | 4419-92-5 |
| Analyte or Component Names | Diethylammonium Salicylate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C7 H6 O3 . C4 H11 N |
| Certifications/Compliance | ISO 17034 |
| Synonym | Benzoic acid, 2-hydroxy-, compd. with N-ethylethanamine (1:1), Salicylic acid, Et2NH salt (6CI), Salicylic acid, compd. with Et2NH (7CI), Salicylic acid, compd. with diethylamine (1:1) (8CI), Diethylamine, compd. with salicylic acid (1:1) (8CI), Ethanamine, N-ethyl-, 2-hydroxybenzoate (9CI), Artrogota, Damsal, Diethylamine salicylate, Diethylammonium salicylate, Salicylate diethylamine, Salicylic acid diethylamine salt |
| InChI Formula | InChI=1S/C7H6O3.C4H11N/c8-6-4-2-1-3-5(6)7(9)10;1-3-5-4-2/h1-4,8H,(H,9,10);5H,3-4H2,1-2H3 |
| SMILES | CCNCC.OC(=O)c1ccccc1O |
| IUPAC Name | N-ethylethanamine;2-hydroxybenzoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction