Learn More
Hamilton™ 250 μL, Model 1725 TLL SYR, Long Life Instrument Syringe
Description
Specification Sheet
- Volume – 250 μL
- Syringe Type – Longlife Gastight
- Syringe Series – 1700
- Termination – PTFE Luer Lock (TLL)
- Plunger Style – Side Post Mounting (XL)
- Fluid Path – Borosilicate glass, PTFE, UHMWPE
- Autoclavable – No
- Gas Sterilizable – Yes
- Pressure – 500 psig (34.5 bar)
- Maximum Temperature – 115°C / 239°F
- Minimum Temperature – 10°C / 50°F
- Barrel Inner Diameter – 0.091 inches (2.30 mm)
- Barrel Outer Diameter – 0.305 inches (7.75 mm)
- Sales Quantity – 1 syringe per pack
Specifications
Specifications
| Product Type | Long Life UHMWPE Syringes, 1700 Series |
| Volume (Metric) | 250 μL |