Learn More
24 Component Mix in Dimethyl Sulfoxide, SPEX CertiPrep™
Product Code. 16320508 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16320508 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16320508 Supplier SPEX Certiprep Supplier No. USPRSC3A
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
1-Butanol71-36-31000μg/mL
1-Pentanol71-41-01000μg/mL
1-Propanol71-23-81000μg/mL
2-Butanol78-92-21000μg/mL
2-Butanone78-93-31000μg/mL
2-Methyl-1-propanol78-83-11000μg/mL
2-Propanol (Isopropanol, Isopropyl alcohol)67-63-01000μg/mL
3-Methyl-1-butanol123-51-31000μg/mL
4-Methyl-2-pentanone108-10-11000μg/mL
Acetone67-64-11000μg/mL
Anisole100-66-31000μg/mL
Butyl acetate123-86-41000μg/mL
Ethanol64-17-51000μg/mL
Ether60-29-71000μg/mL
Ethyl acetate141-78-61000μg/mL
Ethyl formate109-94-41000μg/mL
Isobutyl acetate110-19-01000μg/mL
Isopropyl acetate108-21-41000μg/mL
Isopropylbenzene98-82-81000μg/mL
Methyl acetate79-20-91000μg/mL
n-Heptane142-82-51000μg/mL
n-Pentane109-66-01000μg/mL
Propyl acetate109-60-41000μg/mL
tert-Butyl methyl ether1634-04-41000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Chemical Name or Material 24 Component Mix in Dimethyl Sulfoxide
Name Note USP 467 Standard
Packaging 2mL Amber Prescored Glass Ampule
CAS 71-36-3, 71-41-0, 71-23-8, 78-92-2, 78-93-3
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Explosion Limit 1.8% (volume); 63% (volume)
Ignition Point 270°C
Vapor Pressure 2.5 hPA (20°C)
Analyte or Component Names 1-Butanol; 1-Pentanol; 1-Propanol; 2-Butanol; 2-Butanone; more
Density 1.1g/cm3
Physical Form Fluid
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Element 1-Butanol, 1-Pentanol, 1-Propanol, 2-Butanol, 2-Butanone
Solvent or Matrix Dimethyl Sulfoxide
Quantity 1 mL
Grade USP
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Stock VOA
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: dimethyl sulfoxide,ethanol, isopropylbenzene, 4-methylpentan-2-one, Methyl-tert-butyl ether
Product intended for laboratory use only