missing translation for 'onlineSavingsMsg'
Learn More

24, 25-Dihydroxy VD2, MedChemExpress

Product Code. 16478218
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 16478218

Brand: Medchem Express HY76801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 24, 25-Dihydroxy VD2 is a hydroxylated metabolite of Vitamin D2; a synthetic analog of Vitamin D.

24, 25-Dihydroxy VD2 is a hydroxylated metabolite of Vitamin D2

Specifications

Chemical Name or Material 24, 25-Dihydroxy VD2
CAS 58050-55-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H44O3
Quantity 10 mg
Synonym 24,25-Dihydroxy vitamin D2
Solubility Information DMSO : 100 mg/mL (233.29 mM; Need ultrasonic)
SMILES C=C1/C(C[C@@H](O)CC1)=C\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@@H]([C@H](C)/C=C/C(O)(C)C(C)(C)O)CC3
Molecular Weight (g/mol) 428.65
Formula Weight 428.65
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.