Learn More
Jenway™ 230V Power Supply, EU Plug
Product Code. 11470509
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage unitSize
11470509 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11470509 Supplier Jenway™ Supplier No. 9995601
Only null left
Add to basket
Add to basket
Used with all echem bench models
  • 230V
  • EU plug
  • For use with all echem bench models
Specifications
Type Power Supply, EU Plug
Voltage 230 V